Merck
Cetuximab-induced skin exanthema: prophylactic and reactive skin therapy are equally effective.
Journal of cancer research and clinical oncology (2013-08-07)
Thomas C Wehler, Claudine Graf, Markus Möhler, Jutta Herzog, Martin R Berger, Ines Gockel, Hauke Lang, Matthias Theobald, Peter R Galle, Carl C Schimanski
ABSTRAKT
Treatment with cetuximab is accompanied by the development of an acneiform follicular skin exanthema in more than 80 % of patients. Severe exanthema (grade III/IV) develops in about 9-19 % of patients with the necessity of cetuximab dose reduction or cessation. The study presented was a retrospective analysis of 50 gastrointestinal cancer patients treated with cetuximab in combination with either FOLFIRI or FOLFOX. One cohort of 15 patients received an in-house reactive skin protocol upon development of an exanthema. A second cohort of 15 patients received a skin prophylaxis starting with the first dose of cetuximab before clinical signs of toxicity. A third historic group of 20 patients had received no skin prophylaxis or reactive treatment. 19/20 patients of the historic group developed a skin exanthema. Grade III/IV exanthema was observed six times. Forty percent discontinued cetuximab therapy. The average time to exanthema onset was 14.7 days. Applying the reactive skin protocol after the first occurrence of an exanthema, the exanthema was downgraded as follows: No patients developed grade IV° exanthema, and two patients developed a grade II/III exanthema. In the majority of cases, the reactive skin protocol controlled the exanthema (grade 0-I°). No dose reductions in cetuximab were necessary. Applying the prophylactic skin protocol starting at the beginning of cetuximab application was not superior to the reactive skin protocol. Cetuximab-induced skin exanthema can be coped with a reactive protocol equally effective as compared to a prophylactic skin treatment. A prospective study with higher patient numbers is planned.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Prednicarbate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Metronidazole, BioXtra
Supelco
Phylloquinone (K1), analytical standard
Supelco
Metronidazole, VETRANAL®, analytical standard
Supelco
Metronidazole, analytical standard
Prednicarbate for system suitability A, European Pharmacopoeia (EP) Reference Standard
Phytomenadione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vitamin K1, viscous liquid
Sigma-Aldrich
Minocycline hydrochloride, powder
Supelco
Phytonadione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers