Merck
Cyanide-arene Meisenheimer complex generated in electrospray ionization mass spectrometry using acetonitrile as a solvent.
Journal of the American Society for Mass Spectrometry (2013-08-21)
Barbara Chiavarino, Philippe Maitre, Simonetta Fornarini, Maria Elisa Crestoni
ABSTRAKT
The C - C bond formation activated under negative electrospray ionization of an acetonitrile solution of 1,3,5-trinitrobenzene is reported. The solvent function is to provide a source of cyanide ion, a highly problematic reagent, which is found to attack the electron-deficient aromatic ring to form a covalently bound anionic complex (Meisenheimer complex). The structure of the complex is elucidated by means of collision induced dissociation mass spectrometry and IR multiple photon dissociation spectroscopy in the 'fingerprint' region.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%