Merck
Selective extraction of berberine and palmatine from Huangbai powder.
Pakistan journal of pharmaceutical sciences (2013-09-17)
Cunliu Zhou, Jun Li, Shengjiang Tan, Huiqiong Ye
ABSTRAKT
Berberine and palmatine are two of the main bioactive components in Huangbai, a major Chinese medicinal herb. The current methods to extract these compounds usually involving the usage of inorganic acid and base, are not only complex and time-consuming, but have a low selectivity. In this paper, it was reported that hexane, ethyl acetate and dichloromethane were tested to extract berberine and palmatine from Huangbai powder. The results showed that dichloromethane extracted selectively and effectively berberine and palmatine from Huangbai powder among the examined solvents. In addition, dichloromethane can be recycled and reused, making it a potential candidate for large scale extraction of berberine and palmatine from Huangbai.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Ethyl acetate, analytical standard
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%