- Food for thought: atrioventricular dissociation.
Food for thought: atrioventricular dissociation.
The American journal of medicine (2013-10-15)
Gagan D Singh, Garret B Wong, Jeffrey A Southard, Ezra A Amsterdam
PMID24119821
MATERIAŁY
Numer produktu
Marka
Opis produktu
Lidocaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Lidocaine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Supelco
Lidocaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material