Merck
Functional implications of regulatory B cells in human IgA nephropathy.
Scandinavian journal of immunology (2013-11-14)
Y-Y Wang, L Zhang, P-W Zhao, L Ma, C Li, H-B Zou, Y-F Jiang
ABSTRAKT
IgA nephropathy (IgAN) diagnosis remains largely based upon immunohistologic detection of IgA- and IgG-containing glomerular deposits in renal mesangial cells, and little is known about the underlying pathogenic mechanisms. This study examines the putative contribution of B cell types, including the Breg type, to IgAN pathogenesis. Twenty-four patients with IgAN and proteinuria (Group A: <3.5 g/24 h, n = 13; Group B: >3.5 g/24 h, n = 11) and 10 healthy controls were enrolled. The frequencies of B cell subtypes in venous blood were measured by flow cytometry. Galactose-deficient IgA1 was measurement by ELISA. Needle biopsies were analysed by histology and immunofluorescence microscopy. Correlation between clinical features and B cell subtypes, including the regulatory B (Breg) cells, and Breg cell-derived immunomodulatory cytokine IL-10 was assessed by Spearman's rank correlation test. IgAN patients had significantly higher frequencies of CD27(+) CD19(+) , CD38(+) CD19(+) , CD86(+) CD19(+) and CD5(+) CD19(+) B cells than the healthy controls, but significantly lower levels of Breg cells and intracellular expression of IL-10 protein in the Breg subtype. Serum IgA concentration positively correlated with CD27(+) CD19(+) B cell frequency and negatively correlated with IL-10(+) Breg cell frequency in IgAN patients, and the percentage of CD19(+) CD5(+) CD1d(+) in CD19(+) cells was negatively correlated with the level of serum Gd-IgA1. Furthermore, the frequencies of CD19(+) CD38(+) and CD19(+) CD86(+) in the CD19(+) subpopulation negatively correlated with the estimated glomerular filtration rate of IgAN patients. Several of the CD19(+) B cell subtypes and the IL-10(+) Breg cells are differentially expressed in IgAN patients and may contribute to the disease pathogenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
NADase from porcine brain, ≥0.007 unit/mg solid