Merck
Pregabalin rectifies aberrant brain chemistry, connectivity, and functional response in chronic pain patients.
Anesthesiology (2013-12-18)
Richard E Harris, Vitaly Napadow, John P Huggins, Lynne Pauer, Jieun Kim, Johnson Hampson, Pia C Sundgren, Bradley Foerster, Myria Petrou, Tobias Schmidt-Wilcke, Daniel J Clauw
ABSTRAKT
Chronic pain remains a significant challenge for modern health care as its pathologic mechanisms are largely unknown and preclinical animal models suffer from limitations in assessing this complex subjective experience. However, human brain neuroimaging techniques enable the assessment of functional and neurochemical alterations in patients experiencing chronic pain and how these factors may dynamically change with pharmacologic treatment. To identify the clinical action of pregabalin, a proven analgesic, the authors performed three complementary brain neuroimaging procedures: (proton magnetic resonance spectroscopy, functional magnetic resonance imaging, and functional connectivity magnetic resonance imaging) in 17 chronic pain patients diagnosed with fibromyalgia. The authors found that pregabalin but not placebo reduces combined glutamate + glutamine levels within the posterior insula (pregabalin P = 0.016; placebo P = 0.71). Interestingly, reductions in clinical pain were associated with reductions in brain connectivity of this structure to brain regions within the default mode network during pregabalin (r = 0.82; P = 0.001) but not placebo (r = -0.13; P = 0.63). Response of default mode network regions to experimental pain was also reduced with pregabalin (P = 0.018) but not placebo (P = 0.182). Perhaps most importantly, baseline values for all three neuroimaging markers predicted subsequent analgesic response to pregabalin but not placebo. The results of this study suggest that pregabalin works in part by reducing insular glutamatergic activity, leading to a reduction of the increased functional connectivity seen between brain regions in chronic pain states. The study also supports a role for human brain imaging in the development, assessment, and personalized use of central-acting analgesics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
L-Glutamic acid, FCC
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland