Merck
Initial stages of calcium uptake and mineral deposition in sea urchin embryos.
Proceedings of the National Academy of Sciences of the United States of America (2013-12-18)
Netta Vidavsky, Sefi Addadi, Julia Mahamid, Eyal Shimoni, David Ben-Ezra, Muki Shpigel, Steve Weiner, Lia Addadi
ABSTRAKT
Sea urchin larvae have an endoskeleton consisting of two calcitic spicules. We reconstructed various stages of the formation pathway of calcium carbonate from calcium ions in sea water to mineral deposition and integration into the forming spicules. Monitoring calcium uptake with the fluorescent dye calcein shows that calcium ions first penetrate the embryo and later are deposited intracellularly. Surprisingly, calcium carbonate deposits are distributed widely all over the embryo, including in the primary mesenchyme cells and in the surface epithelial cells. Using cryo-SEM, we show that the intracellular calcium carbonate deposits are contained in vesicles of diameter 0.5-1.5 μm. Using the newly developed airSEM, which allows direct correlation between fluorescence and energy dispersive spectroscopy, we confirmed the presence of solid calcium carbonate in the vesicles. This mineral phase appears as aggregates of 20-30-nm nanospheres, consistent with amorphous calcium carbonate. The aggregates finally are introduced into the spicule compartment, where they integrate into the growing spicule.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis