Merck
Going mobile: AMPA receptors move synapse to synapse in vivo.
Neuron (2013-12-24)
Christopher Rongo
ABSTRAKT
Plasticity models invoke the synaptic delivery of AMPARs, yet we know little about how receptors move in vivo. In this issue of Neuron, Hoerndli et al. (2013) show that lateral diffusion and kinesin-mediated transport move AMPARs between synapses in vivo.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Adenosine 5′-Triphosphatase from porcine cerebral cortex, lyophilized powder, ≥0.3 units/mg protein, pH 7.8