Merck
Mitochondria impact brain function and cognition.
Proceedings of the National Academy of Sciences of the United States of America (2013-12-25)
Martin Picard, Bruce S McEwen
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Estradiol, meets USP testing specifications