- JAMA Pediatrics patient page. Phthalate exposure and health risks.
JAMA Pediatrics patient page. Phthalate exposure and health risks.
JAMA pediatrics (2014-01-08)
Megan Moreno
PMID24394978
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Potassium phthalate monobasic, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland