Merck
Nitecapone and selegiline as effective adjuncts to L-DOPA in reserpine-induced catatonia in mice.
Methods and findings in experimental and clinical pharmacology (2002-05-01)
A Singh, S K Kulkarni
ABSTRAKT
Reserpine-induced catatonia is a widely accepted animal model of Parkinson's disease. In the present study, reserpine (5 mg/kg i.p.) and alpha-methylpara-tyrosine (AMPT) (200 mg/kg i.p.) induced catatonia in mice 20 h and 1 h before the experiment, respectively, as assessed using the rota-rod and bar tests after reserpine treatment. There was a significant decrease in fall-off time in the rota-rod test and a significant increase in time spent on the bar in the bar test as compared to the untreated control mice. Combination therapy with L-DOPA (100 mg/kg i.p.) and carbidopa (10 mg/kg i.p.) was less effective in reversing catatonia as compared to higher doses of L-DOPA (200 mg/kg i.p.) and carbidopa (20 mg/kg i.p.), which showed intense hyperactivity in reserpinized mice. Pretreatment with nitecapone (30 mg/kg i.p.), a COMT inhibitor, or selegiline (10 mg/kg i.p.), a MAO-B inhibitor potentiated the motor stimulant actions of subthreshold doses of the L-DOPA (100 mg/kg i.p.) and carbidopa (10 mg/kg i.p.) combination. Amantadine (40 mg/kg i.p.), but not bromocriptine, potentiated the effects of L-DOPA treatment. The NMDA antagonistic action of amantadine may have beneficial effects. It is concluded that COMT and MAO-B enzymes play an important role in the metabolism of dopamine and administration of a COMT or MAO-B inhibitor may prove to be a better adjunct to L-DOPA therapy than a dopamine receptor agonist in Parkinson's disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nitecapone, ≥98% (HPLC)