Merck
  • Human SMC2 protein, a core subunit of human condensin complex, is a novel transcriptional target of the WNT signaling pathway and a new therapeutic target.
Human SMC2 protein, a core subunit of human condensin complex, is a novel transcriptional target of the WNT signaling pathway and a new therapeutic target.
The Journal of biological chemistry (2012-10-26)
Verónica Dávalos, Lucía Súarez-López, Julio Castaño, Anthea Messent, Ibane Abasolo, Yolanda Fernandez, Angel Guerra-Moreno, Eloy Espín, Manel Armengol, Eva Musulen, Aurelio Ariza, Joan Sayós, Diego Arango, Simó Schwartz
ABSTRAKT
Human SMC2 is part of the condensin complex, which is responsible for tightly packaging replicated genomic DNA prior to segregation into daughter cells. Engagement of the WNT signaling pathway is known to have a mitogenic effect on cells, but relatively little is known about WNT interaction with mitotic structural organizer proteins. In this work, we described the novel transcriptional regulation of SMC2 protein by direct binding of the β-catenin·TCF4 transcription factor to the SMC2 promoter. Furthermore, we identified the precise region in the SMC2 promoter that is required for β-catenin-mediated promoter activation. Finally, we explored the functional significance of down-regulating SMC2 protein in vivo. Treatment of WNT-activated intestinal tumor cells with SMC2 siRNA significantly reduced cell proliferation in nude mice, compared with untreated controls (p = 0.02). Therefore, we propose that WNT signaling can directly activate SMC2 transcription as a key player in the mitotic cell division machinery. Furthermore, SMC2 represents a new target for oncological therapeutic intervention.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Mysz anty-Aktyna (Ab-1) mAb (JLA20), liquid, clone JLA20, Calbiochem®
Sigma-Aldrich
Przeciwciało anty-TCF-4, klon 6H5-3, clone 6H5-3, Upstate®, from mouse
Sigma-Aldrich
Przeciwciało anty-SMC2/hCAP-E, serum, Upstate®