Merck
Effect of bisphosphonates on the rapidly growing male murine skeleton.
Endocrinology (2014-01-16)
Eric D Zhu, Leeann Louis, Daniel J Brooks, Mary L Bouxsein, Marie B Demay
ABSTRAKT
Bisphosphonates are effective for preventing and treating skeletal disorders associated with hyperresorption. Their safety and efficacy has been studied in adults where the growth plate is fused and there is no longitudinal bone growth and little appositional growth. Although bisphosphonate use in the pediatric population was pioneered for compassionate use in the treatment of osteogenesis imperfecta, they are being increasingly used for the treatment and prevention of bone loss in children at risk of hyperresorptive bone loss. However, the effect of these agents on the growing skeleton in disorders other than osteogenesis imperfecta has not been systematically compared. Studies were, therefore, undertaken to examine the consequences of bisphosphonate administration on the growth plate and skeletal microarchitecture during a period of rapid growth. C57Bl6/J male mice were treated from 18 to 38 days of age with vehicle, alendronate, pamidronate, zoledronate, or clodronate at doses selected to replicate those used in humans. Treatment with alendronate, pamidronate, and zoledronate, but not clodronate, led to a decrease in the number of chondrocytes per column in the hypertrophic chondrocyte layer. This was not associated with altered hypertrophic chondrocyte apoptosis or vascular invasion at the growth plate. The effects of pamidronate on trabecular microarchitecture were less beneficial than those of alendronate and zoledronate. Pamidronate did not increase cortical thickness or cortical area/total area relative to control mice. These studies suggest that bisphosphonate administration does not adversely affect skeletal growth. Long-term investigations are required to determine whether the differences observed among the agents examined impact biomechanical integrity of the growing skeleton.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Alendronian sodu, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dichloromethylenediphosphonic acid disodium salt
Sigma-Aldrich
Pamidronate disodium salt hydrate, ≥95% (NMR), solid
Sigma-Aldrich
Alendronate sodium trihydrate, ≥97% (NMR), powder