Merck
  • The effects of tocilizumab on osteitis, synovitis and erosion progression in rheumatoid arthritis: results from the ACT-RAY MRI substudy.
The effects of tocilizumab on osteitis, synovitis and erosion progression in rheumatoid arthritis: results from the ACT-RAY MRI substudy.
Annals of the rheumatic diseases (2014-02-15)
Philip G Conaghan, Charles Peterfy, Ewa Olech, Jeffrey Kaine, David Ridley, Julie Dicarlo, Josh Friedman, Jenny Devenport, Orrin Troum
ABSTRAKT
To examine the imaging-detected mechanism of reduction of structural joint damage progression by tocilizumab (TCZ) in patients with rheumatoid arthritis (RA) using MRI. In a substudy of a randomised, double-blind, phase 3b study (ACT-RAY) of biologic-naïve patients with RA who were methotrexate (MTX)-inadequate responders, 63 patients were randomised to continue MTX or receive placebo (PBO), both in combination with TCZ 8 mg/kg every 4 weeks, with optional additional disease-modifying antirheumatic drugs at week 24 if Disease Activity Score of 28 joints < 3.2. The most symptomatic hand was imaged with 0.2 Tesla extremity MRI at weeks 0, 2, 12 and 52. MR images were scored using Outcome Measures in Rheumatology-Rheumatoid Arthritis Magnetic Resonance Imaging Score. Predictors of week 52 erosion progression were determined by logistic regression analysis. TCZ + PBO (n=32) demonstrated mean improvements in synovitis from baseline to weeks 2 (-0.92; p=0.0011), 12 (-1.86; p<0.0001) and 52 (-3.35; p<0.0001), while TCZ + MTX (n=31) had mean improvements in synovitis at week 12 (-0.88; p=0.0074), but not week 52 (-1.00; p=0.0711). TCZ+PBO demonstrated mean reductions in osteitis at weeks 12 (-5.10; p=0.0022) and 52 (-8.56; p=0.0006), while TCZ+MTX had mean reductions at weeks 2 (-0.21; p<0.05) and 12 (-3.63; p=0.0008), but not week 52 (-2.31; p=0.9749). Mean erosion scores did not worsen in either group. MRI erosion scores at weeks 12 and 52 correlated strongly with radiography erosion scores at week 52 (r>0.80). Baseline synovitis and worsening of osteitis predicted erosion progression. Rapid suppression of synovitis and osteitis with reduction in structural joint damage progression occurred with TCZ, as monotherapy or in combination with MTX, through week 52.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Methotrexate
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard