Merck
Effects of temperature change on cortisol release by common carp Cyprinus carpio.
Journal of fish biology (2014-03-29)
J Jaxion-Harm, F Ladich
ABSTRAKT
Common carp Cyprinus carpio, stressed by fish handling practices, responded with a decrease in cortisol secretion when temperature was lowered from 20 to 14° C within 3·5 h compared to those kept at 20° C.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard