Merck
AROS has a context-dependent effect on SIRT1.
FEBS letters (2014-04-01)
Tarja Kokkola, Tiina Suuronen, Ferdinand Molnár, Juha Määttä, Antero Salminen, Elina M Jarho, Maija Lahtela-Kakkonen
ABSTRAKT
The modulation of protein deacetylase SIRT1 has a vast therapeutic potential in treatment of several aging-associated diseases. Active regulator of SIRT1 (AROS) is a small endogenous protein which was originally reported to activate SIRT1 through a direct interaction in cancer cells. We show that the interaction between the two proteins is weak and does not alter the activity of SIRT1 in non-cancerous human cells. The results of different in vitro SIRT1 activity assays disclosed AROS as an inhibitor of SIRT1. The functional relationship between AROS and SIRT1 proved to be dependent on the biological context and experimental setting.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
Anti-RPS19BP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution