Merck
Impact of drug interactions when medications are stopped: the often forgotten risks.
The Medical journal of Australia (2014-04-08)
Erica Y Tong, Melanie Kowalski, Gary S Yip, Michael J Dooley
MATERIAŁY
Numer produktu
Marka
Opis produktu
Rifampicin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Supelco
Warfarin, PESTANAL®, analytical standard
Supelco
Warfarin, analytical standard