Merck
Force-dependent isomerization kinetics of a highly conserved proline switch modulates the mechanosensing region of filamin.
Proceedings of the National Academy of Sciences of the United States of America (2014-04-08)
Lorenz Rognoni, Tobias Möst, Gabriel Žoldák, Matthias Rief
ABSTRAKT
Proline switches, controlled by cis-trans isomerization, have emerged as a particularly effective regulatory mechanism in a wide range of biological processes. In this study, we use single-molecule mechanical measurements to develop a full kinetic and energetic description of a highly conserved proline switch in the force-sensing domain 20 of human filamin and how prolyl isomerization modulates the force-sensing mechanism. Proline isomerization toggles domain 20 between two conformations. A stable cis conformation with slow unfolding, favoring the autoinhibited closed conformation of filamin's force-sensing domain pair 20-21, and a less stable, uninhibited conformation promoted by the trans form. The data provide detailed insight into the folding mechanisms that underpin the functionality of this binary switch and elucidate its remarkable efficiency in modulating force-sensing, thus combining two previously unconnected regulatory mechanisms, proline switches and mechanosensing.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Proline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
SAFC
L-Proline
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland