Merck
AMP-activated protein kinase regulates intraocular pressure, extracellular matrix, and cytoskeleton in trabecular meshwork.
Investigative ophthalmology & visual science (2014-04-10)
Ayan Chatterjee, Guadalupe Villarreal, Dong-Jin Oh, Min Hyung Kang, Douglas J Rhee
ABSTRAKT
In this study, we investigate how adenosine monophosphate-activated protein kinase (AMPK) affects extracellular matrix (ECM) and cellular tone in the trabecular meshwork (TM), and examine how deletion of its catalytic α2 subunit affects IOP and aqueous humor clearance in mice. Human TM tissue was examined for expression of AMPKα1 and AMPKα2, genomically distinct isoforms of the AMPK catalytic subunit. Primary cultured human TM cells were treated for 24 hours with the AMPK activator 5-amino-1-β-Dffff-ribofuranosyl-imidazole-4-carboxamide (AICAR), under basal or TGF-β2 stimulatory conditions. Conditioned media (CM) was probed for secreted protein acidic and rich in cysteine (SPARC), thrombospondin-1 (TSP-1), and ECM proteins, and cells were stained for F-actin. Cells underwent adenoviral infection with a dominant negative AMPKα subunit (ad.DN.AMPKα) and were similarly analyzed. Intraocular pressure, central corneal thickness (CCT), and aqueous clearance were measured in AMPKα2-null and wild-type (WT) mice. Both AMPKα1 and AMPKα2 are expressed in TM. AICAR activated AMPKα and suppressed the expression of various ECM proteins under basal and TGF-β2 stimulatory conditions. AICAR decreased F-actin staining and increased the phospho-total RhoA ratio (Ser188). Transforming growth factor-β2 transiently dephosphorylated AMPKα. Infection with ad.DN.AMPKα upregulated various ECM proteins, decreased the phospho-total RhoA ratio, and increased F-actin staining. AMPKα2-null mice exhibited 6% higher IOP and decreased aqueous clearance compared with WT mice, without significant differences in CCT or angle morphology. Collectively, our data identify AMPK as a critical regulator of ECM homeostasis and cytoskeletal arrangement in the TM. Mice that are AMPKα2-null exhibit higher IOPs and decreased aqueous clearance than their WT counterparts.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5-Amino-4-imidazolecarboxamide, 95%
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Supelco
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 5′-monophosphate, analytical standard
Dacarbazine impurity B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 5′-monophosphate, ≥93%