Merck
Effect of (131)I gelatin microspheres on hepatocellular carcinoma in nude mice and its distribution after intratumoral injection.
Radiation research (2014-04-12)
Jun-Lin Chi, Chuan-Chao Li, Chuan-Qin Xia, Lin Li, Yu Ma, Jian-Hong Li, Zhuo Chen, Xiao-Li Chen
ABSTRAKT
In this study, we investigated the effect of (131)I gelatin microspheres ((131)I-GMSs) on human hepatocellular carcinoma cells (HepG2) in nude mice (Balb/c) and the biodistribution of (131)I-GMSs after intratumoral injection. The treatment group and control group animals received intratumoral injections of 1 mCi (131)I-GMSs and GMSs unlabeled (131)I, respectively. The size of the implanted tumor was measured once a week for 8 weeks, and the survival time was calculated from the day of injection to 64 days post-injection. Another 35 animals received intratumoral injections of 0.2 mCi (131)I-GMSs and were subject to single-photon emission computed tomography (SPECT) on days 1, 8, 16, 24 and 32 post-injection. Samples of various organs were collected and used to calculate tissue concentrations on days 1, 4, 8, 16 and 24. Free thyroxine (FT4) in fetal bovine serum was tested to evaluate thyroid function. The tumors were collected for histological examination. (131)I-GMSs produced a pronounced reduction in HepG2 tumor volume, and the overall survival was 73.3% in the treatment group and only 13.3% in the control group (P < 0.001). Tissue radioactivity concentration measurements and SPECT demonstrated that the injected (131)I-GMSs mainly accumulated within the tumors. The concentration of FT4 was stable during the observation period. The microspheres could be observed by histological methods on day 32. (131)I-GMSs suppressed the growth of HepG2 in the nude mice and were retained in the tumor for a long period of time after injection. Direct intratumoral injection of (131)I-GMSs offers a promising modality for the treatment of hepatocellular carcinoma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Supelco
L-Thyroxine (T4) solution, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
USP
Lewotyroksyna, United States Pharmacopeia (USP) Reference Standard