Merck
Tazarotene versus tretinoin or adapalene in the treatment of acne vulgaris.
Journal of the American Academy of Dermatology (2000-07-18)
L Kakita
ABSTRAKT
The efficacy and tolerability of tazarotene 0.1% gel in the treatment of acne vulgaris have been compared with those of tretinoin 0.025% gel and adapalene 0.1% gel in multicenter, double-blind, randomized, parallel-group trials. Preliminary results from the tazarotene versus tretinoin trial suggest that once-daily tazarotene is more efficacious than once-daily tretinoin in reducing the numbers of papules and open comedones, and achieves a more rapid reduction in pustules. Both drugs appear to be equally efficacious against closed comedones. Preliminary results from the tazarotene versus adapalene trial suggest that, when tazarotene is applied only half as frequently as adapalene (every other day versus every day), the two drugs achieve comparable reductions in noninflammatory and inflammatory lesion counts. The results from these studies, and a separate split-face tolerability study, suggest that the tolerability of tazarotene gel is clinically comparable to that of tretinoin 0.025% gel, tretinoin 0.1% gel microsphere, and adapalene 0.1% gel.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Adapalene, ≥98% (HPLC)
Adapalene for peak identification, European Pharmacopoeia (EP) Reference Standard
Adapalene, European Pharmacopoeia (EP) Reference Standard
Supelco
Adapalene, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Adapalene, United States Pharmacopeia (USP) Reference Standard
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
Tretinoin, European Pharmacopoeia (EP) Reference Standard
USP
Tretynoina, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Tazarotene, ≥98% (HPLC)