- Efficacy of ofloxacin and other otic preparations for otitis externa.
Efficacy of ofloxacin and other otic preparations for otitis externa.
The Pediatric infectious disease journal (2001-02-15)
R J Ruben
PMID11176588
ABSTRAKT
Ofloxacin otic is as effective in curing otitis externa in children (<12 years of age) as is the preparation combining polymyxin/neomycin plus hydrocortisone and as ciprofloxacin otic. Furthermore ofloxacin otic, as it requires less frequent dosing, is likely to encourage greater patient adherence and consequently to achieve a better cure rate than both of these treatment options. Because a number of cases of otitis externa are complicated by an undiagnosed tympanic membrane perforation, the lack of ototoxicity associated with ofloxacin otic makes it a particularly safe option for use in these patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Ofloxacin, European Pharmacopoeia (EP) Reference Standard