Merck
Use of ampicillin/sulbactam and sultamicillin in pediatric infections: a re-evaluation.
The Journal of international medical research (2001-10-26)
A Dajani
ABSTRAKT
Ampicillin/sulbactam is an effective solution to the emergence of beta-lactamase-mediated resistance among common pediatric pathogens, and is a widely recognized treatment option for a variety of pediatric infections. Recent antimicrobial surveillance data confirm the continued susceptibility of many Gram-positive and Gram-negative aerobes and anaerobes to ampicillin/sulbactam. Pharmacokinetic studies have demonstrated high drug concentrations at a variety of infection sites, including cerebrospinal fluid and bone. Furthermore, clinical studies have shown that ampicillin/sulbactam, administered intravenously, intramuscularly or orally (as the mutual prodrug sultamicillin), is clinically and bacteriologically effective against upper and lower respiratory tract infections, urinary tract infections, skin, bone and soft-tissue infections, and meningitis, and provides effective surgical prophylaxis. Sultamicillin has an excellent tolerability profile, which is associated with a low rate of treatment discontinuation. Accordingly, ampicillin/sulbactam and sultamicillin should be considered first-choice options for the management of a variety of pediatric infections.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Supelco
Ampicillin
Sultamicillin, European Pharmacopoeia (EP) Reference Standard
Sultamicillin for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sulbactam
Sulbactam, European Pharmacopoeia (EP) Reference Standard
Sulbactam for peak identification, European Pharmacopoeia (EP) Reference Standard