Merck
Use of soy protein supplement and resultant need for increased dose of levothyroxine.
Endocrine practice : official journal of the American College of Endocrinology and the American Association of Clinical Endocrinologists (2001-06-26)
D S Bell, F Ovalle
ABSTRAKT
To report a case of difficulty in achieving suppressive serum levels of thyroid hormone because of malabsorption of exogenous levothyroxine attributable to daily ingestion in close temporal relationship to the intake of a soy protein-containing food supplement. We present the relevant history and laboratory data of the current case and provide supportive documentation from the literature. A 45-year-old woman who had hypothyroidism after a near-total thyroidectomy and radioactive iodine ablative therapy for papillary carcinoma of the thyroid required unusually high oral doses of levothyroxine to achieve suppressive serum levels of free thyroxine (T(4)) and thyrotropin (thyroid-stimulating hormone or TSH). She had routinely been taking a "soy cocktail" protein supplement immediately after her levothyroxine. Temporal separation of the intake of the soy protein cocktail from the administration of the levothyroxine resulted in attainment of suppressive serum levels of free T(4) and TSH with use of lower doses of levothyroxine. Administration of levothyroxine concurrently with a soy protein dietary supplement results in decreased absorption of levothyroxine and the need for higher oral doses of levothyroxine to attain therapeutic serum thyroid hormone levels.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)
Supelco
L-Thyroxine (T4) solution, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®
USP
Lewotyroksyna, United States Pharmacopeia (USP) Reference Standard