- Cimetidine: a review of the recent developments and reports in cutaneous medicine.
Cimetidine: a review of the recent developments and reports in cutaneous medicine.
Dermatology online journal (2003-03-18)
Noah Scheinfeld
PMID12639457
ABSTRAKT
Cimetidine, approved by the FDA for inhibition of gastric acid secretion, has been advocated for a number of dermatological diseases. The cutaneous uses and immunological effects of cimetidine have been actively studied over the past few years, and this review summarizes the literature accumulated since 1997.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Cimetidine, European Pharmacopoeia (EP) Reference Standard
Cimetidine for peak identification, European Pharmacopoeia (EP) Reference Standard
Cimetidine for system suitability, European Pharmacopoeia (EP) Reference Standard