Merck
Human health effects of sodium azide exposure: a literature review and analysis.
International journal of toxicology (2003-07-10)
Soju Chang, Steven H Lamm
ABSTRAKT
Sodium azide, used mainly as a preservative in aqueous laboratory reagents and biologic fluids and as a fuel in automobile airbag gas generants, has caused deaths for decades. Its exposure potential for the general population increases as the use of airbags increase. In order to characterize the known health effects of sodium azide in humans and the circumstances of their exposure, the authors conducted a systematic review of the literature from 1927 to 2002 on human exposure to sodium azide and its health effects. The most commonly reported health effect from azide exposure is hypotension, almost independent of route of exposure. Most industrial exposures are by inhalation. Most laboratory exposures or suicide attempts are by ingestion. Most of the reported cases involved persons working in laboratories. The time between exposure and detection of hypotension can predict outcome. Fatal doses occur with exposures of >or=700 mg (10 mg/kg). Nonlethal doses ranged from 0.3 to 150 mg (0.004 to 2 mg/kg). Onset of hypotension within minutes or in less than an hour is indicative of a pharmacological response and a benign course. Hypotension with late onset (>1 hour) constitutes an ominous sign for death. All individuals with hypotension for more than an hour died. Additional health effects included mild complaints of nausea, vomiting, diarrhea, headache, dizziness, temporary loss of vision, palpitation, dyspnea, or temporary loss of consciousness or mental status decrease. More severe symptoms and signs included marked decreased mental status, seizure, coma, arrhythmia, tachypnea, pulmonary edema, metabolic acidosis, and cardiorespiratory arrest. The signs and symptoms from lower exposures (<700 mg) are physiological responses at the vascular level and those at or above are toxicological responses at the metabolic level. There is no specific antidote for sodium azide intoxication. Recommended preventive measures for sodium azide exposure consist of education of people at high risk, such as laboratory workers, regarding its chemical properties and toxicity, better labeling of products containing sodium azide, and strict enforcement of laboratory regulations and access control.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Azydek sodu 0,1 M
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%