Merck
Adefovir dipivoxil: focus on its use in the treatment of chronic hepatitis B.
Reviews in gastroenterological disorders (2005-06-25)
Danial E Baker
ABSTRAKT
Adefovir is classified as a nucleotide reverse transcriptase inhibitor because it acts by inhibiting hepatitis B virus DNA polymerase (reverse transcriptase) and causing DNA chain termination after its incorporation into the viral DNA. Adefovir dipivoxil is indicated for the treatment of chronic hepatitis B in adults with evidence of active viral replication and either evidence of persistent elevations in serum aminotransferases (alanine [ALT] or aspartate [AST]) or histologically active disease. It is useful in the treatment of patients with either hepatitis B e antigen-positive or -negative chronic hepatitis B. The recommended adefovir dipivoxil dose in the treatment of chronic hepatitis B in patients with adequate renal function is 10 mg once daily. Adefovir dipivoxil therapy can reduce viral load, improve ALT, and produce histologic improvement in patients with chronic hepatitis B. Improvements are generally seen within the first few weeks of therapy and have shown persistence up to at least 3 years with continued therapy. Therapy with adefovir dipivoxil is generally well tolerated. However, nephrotoxicity is a risk with adefovir therapy, especially in patients receiving higher doses (30-120 mg/d). Patients should have their renal function monitored closely throughout therapy and may require an adjustment in dose relative to changes in the creatinine clearance. Lactic acidosis and severe hepatomegaly with steatosis may also occur during therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Adefovir dipivoxil