Merck
The mechanisms of action of nicotinamide and zinc in inflammatory skin disease.
Cutis (2006-07-29)
David P Fivenson
ABSTRAKT
Nicotinamide (niacinamide), a physiologically active form of niacin (nicotinic acid), in combination with zinc is being assessed in clinical studies for the treatment of inflammatory skin diseases such as acne vulgaris and bullous pemphigoid. The basis for these investigations is the variety of potential mechanisms of action of nicotinamide and zinc, including an anti-inflammatory effect via inhibition of leukocyte chemotaxis, lysosomal enzyme release, lymphocytic transformation, mast cell degranulation, bacteriostatic effect against Propionibacterium acnes, inhibition of vasoactive amines, preservation of intracellular coenzyme homeostasis, and decreased sebum production. Other possible mechanisms involve suppression of vascular permeability and inflammatory cell accumulation, as well as protection against DNA damage. The goal of this paper is to review the pathophysiology of inflammatory skin diseases and discuss the role, mechanisms of action, and safety of nicotinamide and zinc as therapeutic options for these disorders.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
USP
Niacynamid, United States Pharmacopeia (USP) Reference Standard
Supelco
Nicotinamide (Niacinamide), analytical standard