Merck
[Argon--a biologically active component of atmosphere].
Aviakosmicheskaia i ekologicheskaia meditsina = Aerospace and environmental medicine (2007-04-05)
N B Pavlov
ABSTRAKT
Analysis of the literary and author's own data denies physiological effects of argon in normobaric normoxic atmosphere that could impact human performance, and embryogenesis of lower vertebrates. In hypoxic atmosphere, argon keeps up the normal oxygen consumption by human and rodent organisms, augments the working ability of humans, and exacerbates the hypoxia-driven depression of lower vertebrates embryogenesis. Potential anti-hypoxia benefits from argon introduction into artificial atmosphere are considered.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Argon, ≥99.998%