Merck
Fragrance material review on (-)-alpha-terpineol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-23)
S P Bhatia, C S Letizia, A M Api
ABSTRAKT
A toxicologic and dermatologic review of p-menth-1-en-8-ol (S) when used as a fragrance ingredient is presented.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(−)−α-Terpineol, natural, ≥96%, FCC, FG
Supelco
(+)-α-Terpineol, analytical standard