Merck
Role of salicylic acid in plant abiotic stress.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2008-08-02)
Shu Yuan, Hong-Hui Lin
ABSTRAKT
Salicylic acid (SA) plays many roles in plant physiology. Besides pathogenesis-related resistance, SA is involved in the response to abiotic stress. However, the effects of SA on plant resistance to abiotic stress were found contradictionary, and the actual role of SA in abiotic stress remains unresolved. Generally, deficiency of SA or a very high level of SA increase the plant susceptibility to abiotic stress. The optimal levels for the highest stress tolerance range from 0.1 mM to 0.5 mM for most plants. But the role of SA at a certain level in moderate and severe abiotic stress may be different. This can be attributed to redox regulations in plant cells. In this paper, we discuss the relationship between reactive oxygen species (ROS) and SA, and propose a subsequent intracellular signal transduction network of SA and ROS under abiotic stress. Anti-stress substances besides antioxidant enzymes induced by SA are also summarized.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®