Merck
Reversible phosphorylation of histidine residues in proteins from vertebrates.
Science signaling (2009-03-13)
Susanne Klumpp, Josef Krieglstein
ABSTRAKT
Signaling by kinases and phosphatases that act on serine, threonine, and tyrosine residues of proteins is among the most extensively studied regulatory mechanisms in mammalian cells, and research focused in this area is ongoing. We are just beginning to appreciate that such signaling mechanisms are extended and enriched by the reversible phosphorylation of histidine residues. The most exciting developments in this field to date come from studies on the beta subunit of heterotrimeric guanosine triphosphate-binding proteins (G proteins), the enzyme adenosine 5'-triphosphate-citrate lyase, and now the Ca(2+)-activated K(+) channel KCa3.1, all of which are targeted by nucleoside diphosphate kinase (which phosphorylates histidines) and protein histidine phosphatase (which dephosphorylates phosphorylated histidines).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Histidine
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Histidine, European Pharmacopoeia (EP) Reference Standard