Merck
[Anti-inflammatory effects of pentoxifylline: importance in cardiac surgery].
Der Anaesthesist (2009-11-06)
H V Groesdonk, M Heringlake, H Heinze
ABSTRAKT
Initially introduced as a rheologic agent for use in intermittent claudication due to peripheral artery disease and in ischemic cerebrovascular disease, the methylxanthine derivative pentoxifylline (PTX) has been shown to possess several anti-inflammatory properties which make this drug an interesting immunomodulating adjunct for the management of patients undergoing cardiac surgery. As an unspecific phosphodiesterase inhibitor PTX ameliorates the inflammatory response following a septic stimulus and blunts organ dysfunction after ischemia-reperfusion injury. Apart from this several small clinical studies have shown that the use of PTX may blunt the inflammatory response induced by cardiac surgery using a cardiopulmonary bypass. Additionally it has been shown that the perioperative application of this drug may improve postoperative function of organs at risk, such as the kidneys and liver.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Pentoxifylline, United States Pharmacopeia (USP) Reference Standard
Pentoxifylline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pentoxifylline, solid