Merck
Novel strategies for hydrocortisone replacement.
Best practice & research. Clinical endocrinology & metabolism (2009-06-09)
M Debono, J Newell Price, Richard J Ross
ABSTRAKT
Current therapy with immediate-release hydrocortisone is the most commonly used regimen for replacement in patients with primary and secondary adrenal insufficiency. However, conventional hydrocortisone cannot provide the physiological rhythm of cortisol release. Physicians have used fixed twice- or thrice-daily doses, but these regimens inevitably result in temporary over- or under-replacement. Patients with adrenal insufficiency, although on treatment, have a poor quality of life and an increased mortality. Optimization of current treatment has been attempted with thrice-daily, weight-related dosing, but this still fails to simulate the normal diurnal rhythm of cortisol. Recent research has investigated circadian hydrocortisone therapy imitating the physiological cortisol rhythm. Proof-of-concept studies using hydrocortisone infusions predict improvements in biochemical control and quality of life. Now delayed and sustained release oral formulations of hydrocortisone are being developed, and these offer a more practical and effective solution for patients with adrenal insufficiency and congenital adrenal hyperplasia.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard