- Domperidone-induced acute dystonia and polycystic ovary syndrome.
Domperidone-induced acute dystonia and polycystic ovary syndrome.
Movement disorders : official journal of the Movement Disorder Society (1991-01-01)
U Bonuccelli, A Nocchiero, A Napolitano, A M Paoletti, G B Melis, G U Corsini, A Muratorio
PMID2005928
ABSTRAKT
The occurrence of acute dystonic reactions (ADRs) due to domperidone administration in two young women is reported. In both patients, a typical polycystic ovary (PCO) syndrome was found. The possibility that the relative hyperestrogenism typical of PCO syndrome acted as a facilitating factor for ADR is discussed.