Merck
Inflammatory reactions and hydrocortisone in the setting of cardiac surgery: an overview.
Cardiovascular & hematological agents in medicinal chemistry (2010-11-12)
Jens Heyn, Andres Beiras-Fernandez, Benjamin Luchting, Josef Briegel, Florian Weis
ABSTRAKT
Cardiac surgery with cardiopulmonary bypass (CPB) is associated with activation of the complement system, platelets, neutrophils, monocytes, and macrophages which may lead to systemic inflammatory response syndrome in several cases. Despite modification of surgical techniques, biocompatibility of the bypass circuit and intensive care procedures after operation, CPB is still associated with post-operative morbidity including reduced cardiac function, capillary leak or multi-organ dysfunction. Corticosteroids are known for their anti-inflammatory effects and therefore, they are beneficial in selected trauma or septic patients. Prophylaxis with corticosteroids in cardiac surgery has been used since decades. The studies for methylprednisolone and hydrocortisone, the most commonly used corticosteroids, show conflicting results. For hydrocortisone, which is the mainstream of corticosteroid treatment in septic patients, the number of studies is low, but will increase in the next years. This article reviews the data concerning its use in patients undergoing cardiac surgery, its contraindications, adverse effects, risks, and benefits.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture