Merck
  • Caprylic acid in the effective treatment of intractable medical problems of frequent urination, incontinence, chronic upper respiratory infection, root canalled tooth infection, ALS, etc., caused by asbestos & mixed infections of Candida albicans, Helicobacter pylori & cytomegalovirus with or without other microorganisms & mercury.
Caprylic acid in the effective treatment of intractable medical problems of frequent urination, incontinence, chronic upper respiratory infection, root canalled tooth infection, ALS, etc., caused by asbestos & mixed infections of Candida albicans, Helicobacter pylori & cytomegalovirus with or without other microorganisms & mercury.
Acupuncture & electro-therapeutics research (2011-08-13)
Yoshiaki Omura, Brian O'Young, Marilyn Jones, Andrew Pallos, Harsha Duvvi, Yasuhiro Shimotsuura
ABSTRAKT
There are many causes of frequent urination. Whenever water or fluids are consumed, the patient has to urinate within 10 or 20 min. Often urinary bladder examinations & blood tests show no significant abnormalities, & treatment by anti-bacterial or anti-viral agents does not improve the symptoms significantly. In intractable frequent urination with difficulty holding urine, as well as other intractable medical problems such as frequent coughing, white pus in gingiva, infection of the apex of a root canalled tooth, slow-healing wounds, & ALS, the authors often found coexisting mixed infections of Candida albicans (C.A.), Helicobacter pylori (H.P.), & Cytomegalovirus (CMV) with or without additional bacterial (Chlamydia trachomatis, etc.) or viral infections & increased Asbestos, with or without Hg deposits. We often found various degrees of mixed infections with C.A., H.P., & CMV in the external sphincters of the urethra & in the Trigone of the urinary bladder which consists of (1) a horizontal, band-like area between the 2 ureter openings & (2) the funnel shaped part of the Trigone at the lower half of the urinary bladder. In the coexistence of significant amounts of C.A., H.P. & CMV, the infection cannot be reduced by otherwise effective medicines for H.P. & CMV. However, one optimal dose of Diflucan, or Caprylic acid taken orally or externally applied, rapidly reduced the symptoms significantly. We found the best treatment is to give a combination of an optimal dose of Caprylic acid orally in the form of "CaprilyCare" or "Caprylic Acid," with a capsule of Omega-3 Fish Oil as an anti-viral agent, Amoxicillin, Substance Z & a Cilantro tablet. We found that an optimal dose of Caprylic acid increases normal cell telomere (NCT) to a desirable 750 ng BDORT units while Diflucan increases NCT by only 25 ng BDORT units, & with Omega-3 fish oil, leads to a mutual cancellation of both drugs. Thus, Caprylic acid is superior to & less expensive than Diflucan, & has potential application for anti-cancer, anti-aging, anti-Alzheimer's disease, anti-Autism, anti-infection, & general circulatory improvement.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Caprylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Caprylic Acid (Octanoic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Octanoic acid, analytical standard
Sigma-Aldrich
Octanoic acid, ≥99%
Sigma-Aldrich
Octanoic acid, natural, ≥98%, FG
Sigma-Aldrich
Octanoic acid, ≥98%
Sigma-Aldrich
Octanoic acid, ≥98%, FG