Merck
Staurosporine: a prototype of a novel class of inhibitors of tumor cell invasion?
Journal of the National Cancer Institute (1990-11-21)
C A O'Brian, N E Ward
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Staurosporine, VETRANAL®, analytical standard
Sigma-Aldrich
Staurosporine from Streptomyces sp., Molecular Biology, ≥95% (HPLC)
Sigma-Aldrich
Staurosporine from Streptomyces sp., ≥98% (HPLC), film
Sigma-Aldrich
Staurosporyna, Staurosporine Streptomyces is a potent, cell-permeable, reversible, ATP-competitive and broad spectrum inhibitor of protein kinases.