- Valproic acid-induced eosinophilic pleural effusion: a case report and review of the literature.
Valproic acid-induced eosinophilic pleural effusion: a case report and review of the literature.
The neurologist (2012-01-06)
Zvi Kamenetsky, Nael Da'as, Yaacov Esayag, Yosef Kleinman, Noah Samuels
PMID22217614
ABSTRAKT
We present a case of eosinophilic pleural effusion due to valproic acid (VPA), a rare adverse event that has been reported previously. A 30-year-old male patient presented with respiratory symptoms and right-sided pleuritic pain, within days of initiation of VPA treatment for a generalized seizure. Chest radiography revealed a moderate-sized right-sided pleural effusion, which was an exudate with a pH of 7.54 and 48% eosinophils. Symptoms resolved soon after discontinuation of the drug, as did the effusion in a repeat radiograph 3 weeks later. The likelihood that VPA was the cause of the effusion is examined.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Valproic acid, European Pharmacopoeia (EP) Reference Standard