Merck
Nicotinamide in dermatology and photoprotection.
Skinmed (2012-01-20)
Devita Surjana, Diona L Damian
ABSTRAKT
Nicotinamide (the amide form of vitamin B3) has been used in dermatology for more than 40 years for a diverse range of conditions including acne, rosacea, autoimmune bullous dermatoses, and now the treatment and prevention of photoaging and photoimmunosuppression. The broad clinical effects of nicotinamide may be explained by its role as a cellular energy precursor, a modulator of inflammatory cytokines, and an inhibitor of the nuclear enzyme poly(adenosine diphosphate-ribose) polymerase-1, which plays a significant role in DNA repair, maintenance ofgenomic stability, and cellular response to injury including inflammation and apoptosis. This review outlines the use of nicotinamide for inflammatory dermatoses and photoaging and focuses on its emerging role in photoprotection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Niacynamid, United States Pharmacopeia (USP) Reference Standard
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Nicotinamide (Niacinamide), analytical standard