Merck
Cholecalciferol.
Topics in companion animal medicine (2013-06-26)
Michael E Peterson, Kerstin Fluegeman
ABSTRAKT
The primary source of exposure to cholecalciferol in dogs and cats is ingestion of rodenticide baits with vitamin D3 as the active ingredient. Other sources of this toxin are human medications and rarely, contaminated pet food. Although the reported lethal dose 50% for cholecalciferol is 88 mg/kg, deaths have been seen with an individual exposure of 2 mc g/kg in dogs. Clinical signs are induced by profound hypercalcemia affecting multiple body systems. Clinical presentations may include anorexia, depression, muscle weakness, vomiting, polyuria, polydipsia, dehydration, abdominal pain, hematemesis, melena, and bradycardia. Tissue mineralization may develop if calcium × phosphorous product is greater than 60. Serum testing for hypercalcemia, hyperphosphatemia, and decreased serum parathyroid hormone are confirmatory. Initial treatment relies upon decontamination with emesis induction followed by administration of pulse-dose activated charcoal designed to interfere with the extensive enterohepatic recirculation of toxin. Medical management is designed to decrease serum calcium levels by use of intravenous fluid diuresis with administration of furosemide and prednisolone. Biphosphate pamidronate is used to inhibit calcium release from the bone. Phosphate binders aid in decreasing phosphate availability to interact with calcium. The prognosis is better if treatment is instituted early before development of hypercalcemia and hyperphosphatemia enables tissue mineralization to progress.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Supelco
Cholecalciferol (D3), analytical standard
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®