Merck
Glycine 1.5% for irrigation should be abandoned.
Urologia internationalis (2013-09-21)
Robert G Hahn
ABSTRAKT
Glycine 1.5% has long maintained a dominating role as an irrigating solution for monopolar transurethral resection of the prostate (TURP), as well as for certain other transurethral procedures. This review summarizes the findings of systematic experimental and clinical studies in which glycine 1.5% for irrigation was infused/absorbed and the outcome compared to at least one other irrigating fluid, including the isotonic saline used for bipolar TURP. There were 11 studies in animals, 3 in volunteers and 6 in patients undergoing TURP. With only one exception, which is probably due to low power, these studies either show a poorer outcome after administration or absorption of glycine solution or else that glycine 2.2% is more toxic than glycine 1.5%. The poorer outcomes consisted of more tissue damage or higher mortality (animals) or more symptoms (volunteers and patients). The safety of monopolar TURP would be improved by replacing glycine 1.5% with some other electrolyte-free fluid. The author argues that glycine 1.5% should be abandoned completely.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine 1 M solution