Merck
[Protein determination by binding with the dye Coomassie brilliant blue G-250].
Biokhimiia (Moscow, Russia) (1994-06-01)
V S Gasparov, V G Degtiar'
ABSTRAKT
Possible applications of the Coomassie brilliant blue G-250 dye method for protein assay are reviewed. The physico-chemical and optical properties of the dye are considered in detail. The data on the protein-binding mechanism are generalized. Modifications of the Coomassie blue G-250 quantitative protein assays are critically considered. The applications of the method in medicine, biochemistry and biotechnology are discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Brilliant Blue G solution, Concentrate
Sigma-Aldrich
Brilliant Blue G, pure
Sigma-Aldrich
Brilliant Blue G, 250, suitable for microscopy