Merck
Increases in rates of resistance to trimethoprim.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (1997-01-01)
P Huovinen
ABSTRAKT
Trimethoprim alone or in combination with a sulfonamide is an effective and relatively inexpensive antibacterial medication. However, a dramatic increase in the rate of resistance to trimethoprim along with high-level resistance to sulfonamides has been seen during the past two decades. The mechanisms of resistance show a remarkable evolutionary adaptation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Trimethoprim, United States Pharmacopeia (USP) Reference Standard
Supelco
Trimethoprim, VETRANAL®, analytical standard
Trimethoprim, European Pharmacopoeia (EP) Reference Standard