Merck
5'-Nucleotidase: molecular structure and functional aspects.
The Biochemical journal (1992-07-15)
H Zimmermann
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5′-Nucleotidase human, recombinant, expressed in CHO cells, vial of 6-12 μg