Merck
  • Non-biologic remission maintenance therapy in adult patients with ANCA-associated vasculitis: a systematic review and network meta-analysis.
Non-biologic remission maintenance therapy in adult patients with ANCA-associated vasculitis: a systematic review and network meta-analysis.
Joint, bone, spine : revue du rhumatisme (2014-01-07)
Glen S Hazlewood, Claudia Metzler, George A Tomlinson, Wolfgang L Gross, Brian M Feldman, Loic Guillevin, Christian Pagnoux
ABSTRAKT
To determine the comparative efficacy of non-biologic treatments for remission maintenance in ANCA-associated vasculitis. We identified all randomized trials comparing leflunomide, azathioprine, methotrexate or mycophenolate mofetil in adult patients with granulomatosis with polyangiitis or microscopic polyangiitis. Relapse-free survival was compared through hazard ratios (HR) using a Bayesian fixed-effects network meta-analysis. Multiple sensitivity analyses were performed to explore biases identified in one trial using original trial data. Three trials were available (leflunomide-methotrexate, methotrexate- azathioprine, azathioprine-mycophenolate). Mycophenolate was inferior to all treatments, although the 95% credible interval (CrI) of the HR relative to methotrexate crossed 1. Leflunomide was superior to azathioprine (HR 0.43 [95% CrI: 0.14-1.3]) and methotrexate (HR 0.47 [95% CrI: 0.18-1.2]), although the 95% CrI also crossed 1. There was a 90% probability that leflunomide was the best treatment. After down weighting the effect of leflunomide vs. methotrexate for early trial termination and slow MTX dose escalation, there remained a 55% probability leflunomide was best. Based on indirect evidence, leflunomide is effective in maintaining remission in granulomatosis with polyangiitis or microscopic polyangiitis relative to other non-biologic treatments. Further randomized trials of leflunomide are needed for confirmation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Mycophenolic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Azathioprine, ≥98%
Mycophenolate mofetil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
USP
Mycophenolate mofetil, United States Pharmacopeia (USP) Reference Standard
Azathioprine, European Pharmacopoeia (EP) Reference Standard
Supelco
Azathioprine, Pharmaceutical Secondary Standard; Certified Reference Material
Leflunomide, European Pharmacopoeia (EP) Reference Standard
USP
Leflunomide, United States Pharmacopeia (USP) Reference Standard
Leflunomide for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture
Supelco
Mycophenolic acid, analytical standard
Sigma-Aldrich
Leflunomide, Immunosuppressant
Sigma-Aldrich
Mycophenolic acid, ≥98%
Mycophenolate mofetil for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Leflunomide, Pharmaceutical Secondary Standard; Certified Reference Material