Merck
Hemopexin-dependent heme uptake via endocytosis regulates the Bach1 transcription repressor and heme oxygenase gene activation.
Biochimica et biophysica acta (2014-03-13)
Hiroshi Hada, Takuma Shiraki, Miki Watanabe-Matsui, Kazuhiko Igarashi
ABSTRAKT
Intracellular heme plays versatile roles in a variety of physiological processes including mitochondrial respiration. Heme also induces the expression of genes such as heme oxygenase-1 (HO-1) by inactivating the transcription repressor Bach1 through direct binding. However, the source of heme for the regulation of the Bach1-HO-1 axis has been unclear. Considering that extracellular heme exists as a complex with hemopexin (Hx) in serum under the physiological conditions, heme-Hx complex may deliver heme for the gene regulation. Using a mammalian expression system, high secretory recombinant Hx (rHx) was developed. We examined the effects of rHx-bound heme on HO-1 expression and Bach1 in Hepa-1c1c7 liver cells and THP-1 macrophage cells. We investigated the uptake pathway of rHx-bound heme by treating cells with chlorpromazine (CPZ). rHx-bound heme induced the expression of HO-1 and decreased the level of Bach1 protein. CPZ inhibited the induction of the HO-1 expression by rHx-bound heme. rHx-bound heme was internalized into the cells via endocytosis, resulting in HO-1 expression and inactivation of Bach1. The Bach1-dependent repression of the HO-1 expression is under the control of the Hx-dependent uptake of extracellular heme. Heme may regulate Bach1 as an extracellular signaling molecule.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Supelco
Chlorpromazine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hemopexin from human plasma, ≥95% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.