Merck
Effects of biochar and the geophagous earthworm Metaphire guillelmi on fate of (14)C-catechol in an agricultural soil.
Chemosphere (2014-05-31)
Jun Shan, Yongfeng Wang, Jianqiang Gu, Wenqiang Zhou, Rong Ji, Xiaoyuan Yan
ABSTRAKT
Both biochar and earthworms can exert influence on behaviors of soil-borne monomeric phenols in soil; however, little was known about the combined effects of biochar and earthworm activities on fate of these chemicals in soil. Using (14)C-catechol as a representative, the mineralization, transformation and residue distribution of phenolic humus monomer in soil amended with different amounts of biochar (0%, 0.05%, 0.5%, and 5%) without/with the geophagous earthworm Metaphire guillelmi were investigated. The results showed biochar at amendment rate <0.5% did not affect (14)C-catechol mineralization, whereas 5% biochar amendment significantly inhibited the mineralization. Earthworms did not affect the mineralization of (14)C-catechol in soil amended with <0.5% biochar, but significantly enhanced the mineralization in 5% biochar amended soil when they were present in soil for 9 d. When earthworms were removed from the soil, the mineralization of (14)C-catechol was significantly lower than that of in earthworm-free soil indicating that (14)C-catecholic residues were stabilized during their passage through earthworm gut. The assimilation of (14)C by earthworms was low (1.2%), and was significantly enhanced by biochar amendment, which was attributed to the release of biochar-associated (14)C-catecholic residues during gut passage of earthworm.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,2-dihydroksybenzen, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%