Merck
  • Enhanced performance of a three-zone simulated moving bed chromatography for separation of succinic acid and lactic acid by simultaneous use of port-location rearrangement and partial-feeding.
Enhanced performance of a three-zone simulated moving bed chromatography for separation of succinic acid and lactic acid by simultaneous use of port-location rearrangement and partial-feeding.
Journal of chromatography. A (2014-06-03)
Sungyong Mun
ABSTRAKT
The performance of a three-zone simulated moving bed (SMB) chromatographic process for separation of succinic acid and lactic acid has been improved to a certain extent in previous researches by applying either a partial-feeding (PF) or a port-location rearrangement (PR) to its operation. To make a further improvement, the strategy of applying both PF and PR simultaneously to the three-zone SMB operation was proposed in this study. The results from both equilibrium-theory analysis and detailed simulation proved that the proposed strategy, which was called PF-PR in this article, had the benefit of a synergy between the individual merits of PF and PR in the three-zone SMB performance. As a consequence, the PF-PR mode could surpass the PF and the PR modes by a wide margin and the classical mode by a dramatic margin in the aspects of separation performance and throughput.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Supelco
Succinic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Kwas mlekowy, United States Pharmacopeia (USP) Reference Standard
Supelco
Succinic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactic acid, 88%, FCC
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material