Merck
Determining better treatments for neurocysticercosis.
The Lancet. Infectious diseases (2014-07-08)
Gagandeep Singh, Arthur Clinton White
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Praziquantel, VETRANAL®, analytical standard
Supelco
Praziquantel, Pharmaceutical Secondary Standard; Certified Reference Material
Praziquantel, European Pharmacopoeia (EP) Reference Standard
Albendazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Albendazole, analytical standard, ≥98%
Praziquantel for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Albendazole, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Albendazole, United States Pharmacopeia (USP) Reference Standard